Masterflex® L/S® Tang Adapter Kit, 2 Large and 2 Small Tang Adapters
Katalogové číslo MFLX07519-01
Dodavatel: Avantor Fluid Handling
Specifikace
- Pump series:L/S
- Popis:L/S Tang Adapter Kit, 2 large and 2 small per kit
- Bal.:1
Specifikace
O tomto produktu
Extend the life of your Masterflex pump head with these replacement parts.
- Suitable for use with PTFE Diaphragm pump head and Multichannel Cartridge pump heads
Replacement parts to keep your pump head operating even longer. Tang adapter kits provides a tight tang shaft connection and reduces noise. Use large adapter for 75W (1/10 hp) drives and small adapter for 37W (1/20 hp drives).
- Pump series:L/S
- Popis:L/S Tang Adapter Kit, 2 large and 2 small per kit
- Environmentally Preferable:
- Bal.:1