Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Non-DEHP, Avantor®
Dodavatel: Avantor Fluid Handling
Certifikáty
O tomto produktu
High resistance to kinking & collapse.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.
Certifications: Resin meets USP Class VI requirements, non-DEHP.
Temperature range: -31 to 185 °F (-35 to 85 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
- Environmentally Preferable:
Specifikace
- Klarita:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 152 m (500 ft.)
- 152 m (500 ft.)
- 152 m (500 ft.)
- 152 m (500 ft.)
- 152 m (500 ft.)
- Maximální teplota:
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- Minimální teplota:
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- Regulatory approvals:
- Non-DEHP;USP
- Non-DEHP;USP
- Non-DEHP;USP
- Non-DEHP;USP
- Non-DEHP;USP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes
- Yes
- Yes
- Yes
- Yes