Water Pump, 6 strokes/gallon, 3 foot inlet tube
Catalog # MFLX70607-14
Supplier: Avantor Fluid Handling
Specifications
- Průtoková rychlost:6 strokes/gal.
- Popis:Water Pump, 6 strokes/gallon, 3 foot inlet tube
- Máčené materiály:PVC body, hose and shaft
- Tube length, inlet:36"
- Záruka:1 Month
- Bal.:1
Specifications
About this item
Ideal for removing water in municipal and irrigation markets in addition to facilities maintenance.
- Recommended for aqueous solutions
- Withstand temperatures up to 120F (49C)
- Self-priming, will not siphon
This hand-operated drum pump is ideal for pumping water and aqueous solutions. Constructed with a PVC body, hose and shaft, this pump offers 6 strokes/gallon.
- Průtoková rychlost:6 strokes/gal.
- Popis:Water Pump, 6 strokes/gallon, 3 foot inlet tube
- Environmentally Preferable:
- Máčené materiály:PVC body, hose and shaft
- Tube length, inlet:36"
- Záruka:1 Month
- Bal.:1