Masterflex® Direct Lift 2- and 3-way Solenoid Valves, Avantor®
Ideal for use with high-purity or corrosive fluids.
- All wetted parts made of inert PTFE
- Minimal dead volume reduces problems with trapped liquids
Rated for 26" Hg vacuum. Maximum fluid temperature is 158 °F (70 °C). All valves have 15" lead wires. Valves are compact and lightweight — mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.
- Environmentally Preferable:
- Catalog No:
- MFLX01540-10
- MFLX01540-14
- MFLX01540-01
- MFLX01540-17
- MFLX01540-02
- MFLX01540-18
- MFLX01540-04
- MFLX01540-08
- MFLX01540-09
- MFLX01540-11
- MFLX01540-12
- MFLX01540-13
- Model:
- 01540-10
- 01540-14
- 01540-01
- 01540-17
- 01540-02
- 01540-18
- 01540-04
- 01540-08
- 01540-09
- 01540-11
- 01540-12
- 01540-13
- Maximální tlak:
- 2 bar (30 psi)
- 4,1 bar (60 psi)
- 30 psi
- 1,2 bar (18 psi)
- 2 bar (30 psi)
- 1,2 bar (15 psi)
- 4,1 bar (60 psi)
- 1,2 bar (18 psi)
- 2 bar (30 psi)
- 2 bar (30 psi)
- 2 bar (30 psi)
- 4,1 bar (60 psi)
- Maximální teplota:
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- Vstupní otvor:
- 1,5875 mm (0,0625")
- 1,5875 mm (0,0625")
- 1,5875 mm (0,0625")
- 3,96748 mm (0,1562")
- 1,5875 mm (0,0625")
- 3,96748 mm (0,1562")
- 1,5875 mm (0,0625")
- 3,96748 mm (0,1562")
- 1,5875 mm (0,0625")
- 1,5875 mm (0,0625")
- 1,5875 mm (0,0625")
- 1,5875 mm (0,0625")
- Výkon:
- 24 VDC, 4 W
- 24 VDC, 8 W
- 12 VDC, 4 W
- 12 VDC, 8 W
- 24 VDC, 4 W
- 24 VDC, 8 W
- 24 VDC, 8 W
- 24 VDC, 8 W
- 12 VDC, 4 W
- 12 VDC, 4 W
- 24 VDC, 4 W
- 12 VDC, 8 W
- Čas odezvy:
- 20 ms
- 25 ms
- 20 ms
- 30 ms
- 20 ms
- 30 ms
- 25 ms
- 30 ms
- 20 ms
- 20 ms
- 20 ms
- 25 ms
- Máčené materiály:
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- Záruka:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Frequently Bought Together