Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
Katalogové číslo MFLX30544-20
Dodavatel: Avantor Fluid Handling
Specifikace
- Sanitary clamp size (Port 1):3"
- Popis:Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
- Bal.:1
- Fitting material:Platinum-Cured Silicone
- Sanitary gasket size:3"
- Záruka:1 year
Specifikace
O tomto produktu
Easily sample fluid in critical high-purity system.
- Three internal ports accept thermocouple probe0
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.
- Sanitary clamp size (Port 1):3"
- Popis:Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
- Environmentally Preferable:
- Bal.:1
- Fitting material:Platinum-Cured Silicone
- Sanitary gasket size:3"
- Záruka:1 year