Masterflex® Hand Tool for Securing Plastic Hose Clamps
Catalog # MFLX06832-97
Supplier: Avantor Fluid Handling
Specifications
- Popis:Hand Tool for Securing Plastic Hose Clamps
- Bal.:1
- Min. int.Ø:6,8 mm (0,269")
Specifications
About this item
Ideal for securing plastic hose clamps.
- Ensures secure fit
Use the hand tool with plastic hose clamps for easy clamp installation.
- Popis:Hand Tool for Securing Plastic Hose Clamps
- Environmentally Preferable:
- Bal.:1
- Min. int.Ø:6,8 mm (0,269")