Masterflex® Flow Indicators for Liquids, Avantor®
O tomto produktu
Easily view flow through clear plastic lens.
- Indicators feature a bright orange rotor that spins to provide visual verification
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.
8 Dostupných variant níže
Product Details & Documents
Easily view flow through clear plastic lens.
- Indicators feature a bright orange rotor that spins to provide visual verification
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.
Specifications for Product Family
Specifikace
- Catalog No:
- MFLX32779-06
- MFLX32779-04
- MFLX32779-09
- MFLX32779-08
- MFLX32779-02
- MFLX32779-10
- MFLX32779-00
- MFLX32880-32
- Model:
- 32779-06
- 32779-04
- 32779-09
- 32779-08
- 32779-02
- 32779-10
- 32779-00
- 32880-32
- Min. provozní teplota:
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- Maximální provozní teplota:
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- Máčené materiály:
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- O-Ring material:
- Viton®
- Viton®
- Viton®
- Viton®
- Buna-N
- Buna-N
- Typ zobrazení:
- High Visibility Orange, Molded Nylon Rotor
- High Visibility Orange, Molded Nylon Rotor
- High Visibility Orange, Molded Nylon Rotor
- High Visibility Orange, Molded Nylon Rotor
- High Visibility Orange, Molded Nylon Rotor
- Záruka:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifikace
Product Family Options
Informace o produktu
- SekvenceHousing materialMaximální průtokPipe sizeDostupnostPrice
- MFLX32779-06Flow Indicator; Brass, 1/2" NPT(F), Orange RotorFlow Indicator; Brass, 1/2" NPT(F), Orange Rotor01Brass20 gal./min1/2" NPT(F)Přihlásit se pro dostupnost1 * 1 KS8 230,00 KčSpecifikace:1 year warranty
More
Specifications
Cat. No.MFLX32779-06Model32779-06Min. provozní teplota–29 °C (–20 °F)Maximální provozní teplota100 °C (212 °F)Máčené materiályRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Typ zobrazeníHigh Visibility Orange, Molded Nylon RotorZáruka1 year1 year warranty - MFLX32779-04Flow Indicator; Brass, 1/4" NPT(F), Orange RotorFlow Indicator; Brass, 1/4" NPT(F), Orange Rotor02Brass5 gal./min1/4" NPT(F)Přihlásit se pro dostupnost1 * 1 KS8 230,00 KčSpecifikace:1 year warranty
More
Specifications
Cat. No.MFLX32779-04Model32779-04Min. provozní teplota–29 °C (–20 °F)Maximální provozní teplota100 °C (212 °F)Máčené materiályRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Typ zobrazeníHigh Visibility Orange, Molded Nylon RotorZáruka1 year1 year warranty - MFLX32779-09Flow Indicator; Brass, 1" NPT(F)Flow Indicator; Brass, 1" NPT(F)03Brass30 gal./min1" NPT(F)Přihlásit se pro dostupnost1 * 1 KS8 250,00 KčSpecifikace:1 year warranty
More
Specifications
Cat. No.MFLX32779-09Model32779-09Min. provozní teplotaMaximální provozní teplota100 °C (212 °F)Máčené materiályRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Typ zobrazeníZáruka1 year1 year warranty - MFLX32779-08Flow Indicator; Brass, 3/4" NPT(F), Orange RotorFlow Indicator; Brass, 3/4" NPT(F), Orange Rotor04Brass30 gal./min3/4" NPT(F)Přihlásit se pro dostupnost1 * 1 KS8 250,00 KčSpecifikace:1 year warranty
More
Specifications
Cat. No.MFLX32779-08Model32779-08Min. provozní teplota–29 °C (–20 °F)Maximální provozní teplota100 °C (212 °F)Máčené materiályRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Typ zobrazeníHigh Visibility Orange, Molded Nylon RotorZáruka1 year1 year warranty - MFLX32779-02Flow Indicator; Polypropylene, 1/2" NPT(F), Orange RotorFlow Indicator; Polypropylene, 1/2" NPT(F), Orange Rotor05PP20 gal./min1/2" NPT(F)Přihlásit se pro dostupnost1 * 1 KS4 030,00 KčSpecifikace:1 year warranty
More
Specifications
Cat. No.MFLX32779-02Model32779-02Min. provozní teplota–29 °C (–20 °F)Maximální provozní teplota82 °C (180 °F)Máčené materiályRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialBuna-NTyp zobrazeníHigh Visibility Orange, Molded Nylon RotorZáruka1 year1 year warranty - MFLX32779-10Flow Indicator; Polypropylene, 1/4" NPT(F)Flow Indicator; Polypropylene, 1/4" NPT(F)065 gal./min1/4" NPT(F)Přihlásit se pro dostupnost1 * 1 KS6 650,00 KčSpecifikace:CE CompliantUL recognized
More
Specifications
Cat. No.MFLX32779-10Model32779-10Min. provozní teplotaMaximální provozní teplota82 °C (180 °F)Máčené materiályO-Ring materialTyp zobrazeníZáruka1 yearCE CompliantUL recognized - MFLX32779-00Flow Indicator; Polypropylene, 1/4" NPT(F), Orange RotorFlow Indicator; Polypropylene, 1/4" NPT(F), Orange Rotor07PP5 gal./min1/4" NPT(F)Přihlásit se pro dostupnost1 * 1 KS4 030,00 KčSpecifikace:1 year warrantyCE Compliant
More
Specifications
Cat. No.MFLX32779-00Model32779-00Min. provozní teplota–29 °C (–20 °F)Maximální provozní teplota82 °C (180 °F)Máčené materiályRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialBuna-NTyp zobrazeníHigh Visibility Orange, Molded Nylon RotorZáruka1 year1 year warrantyCE Compliant - MFLX32880-32Flow Sensor/Switch; Polypropylene, 1/2" NPT(F), 24 VDCFlow Sensor/Switch; Polypropylene, 1/2" NPT(F), 24 VDC0820 gal./min1/2" NPT(F)Přihlásit se pro dostupnost1 * 1 KS14 580,00 KčSpecifikace:
More
Specifications
Cat. No.MFLX32880-32Model32880-32Min. provozní teplotaMaximální provozní teplota82 °C (180 °F)Máčené materiályO-Ring materialTyp zobrazeníZáruka1 year