Masterflex® L/S® High-Performance Pump Head Rotor Assembly Clip
Catalog # MFLX77250-69
Supplier: Avantor Fluid Handling
Specifications
- Pump series:L/S
- Popis:L/S High-Performance Rotor Assembly Clip for 77250-62 /-82
- Záruka:2 years
- Bal.:1
Specifications
About this item
Get the most out of your Masterflex® L/S® High-Performance Pump Head with these replacement parts.
- Suitable for use with High-Performance pump heads 77250-62 and -82
Replacement parts to keep your pump head operating even longer.
- Pump series:L/S
- Popis:L/S High-Performance Rotor Assembly Clip for 77250-62 /-82
- Environmentally Preferable:
- Záruka:2 years
- Bal.:1