0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Média kultivační
CMRL 1066, supplemented, Corning®
CMRL 1066, supplemented, Corning®
Catalog # MDTC99-603-CV
Supplier:
Corning
CAS Number:
Expand image
CMRL 1066, supplemented, Corning®
Catalog # MDTC99-603-CV
Supplier:
Corning
CAS Number:
Specifications
Bal.:
500 mL
Popis:
CMRL 1066, supplemented, storage temperature: 2 to 8 °C
Specifications
About this item
Product Details
Documentation
Product Details
Documentation