CPC® Miniature Quick-Disconnect Fittings, Hose Barb Complete Couplings
Dodavatel: Avantor Fluid Handling
Certifikáty
O tomto produktu
No more kinked tubing - allows tubing to rotate freely.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
- Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle ¹/₁₆" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
The complete couplings come with one insert and one body fitting.
- Environmentally Preferable:
Specifikace
- Catalog No:
- MFLX06363-91BLK
- MFLX06363-88BLK
- MFLX06363-92BLK
- MFLX06363-88
- MFLX06363-91
- MFLX06363-89BLK
- MFLX06363-93BLK
- MFLX06363-93
- Maximální teplota:
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- Minimální teplota:
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- O-Ring material:
- Buna N
- Buna N
- Buna N
- Buna N
- Buna N
- EPDM
- EPDM
- EPDM
- Sterilizace:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Valve material:
- None
- None
- Acetal
- None
- None
- None
- None
- None
- Valve spring material:
- 316 SS
- Záruka:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year