Masterflex® Transfer Tubing, Versilon™ C-544-A IB, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
No plasticizers.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
The Versilon™ C-544-A IB ether-based polyurethane pressure tubing, is ideal for abrasive and viscous slurry transfer, instrumentation control lines, food and cosmetic processing, and coolant recovery systems. Characteristics include exceptional abrasion and tear resistance; excellent resistance to oils, grease and fuels. The transparent tubing features plasticizer-free construction and has a temperature range of -100 to 180 °F (-73.3 to 82.2 °C). It is FDA, NSF 51, and NSF 61 compliant. Not recommended for sterilization.
- Environmentally Preferable:
Specifications
- Model:
- 95637-06
- 95637-07
- 95637-05
- 95637-01
- 95637-04
- 95637-02
- 95637-00
- Klarita:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- Maximální tlak:
- 6,6 bar (95 psi)
- 5,5 bar (80 psi)
- 8,3 bar (120 psi)
- 14,1 bar (205 psi)
- 10,3 bar (150 psi)
- 13,4 bar (195 psi)
- 19 bar (275 psi)
- Maximální teplota:
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- Minimální teplota:
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- –73 °C (–100 °F)
- Regulatory approvals:
- FDA;NSF-61
- FDA;NSF-61
- FDA;NSF-61
- FDA;NSF-61
- FDA;NSF-61
- FDA;NSF-61
- FDA;NSF-61
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- Pracovní tlak:
- 6,6 bar (95 psi)
- 5,5 bar (80 psi)
- 8,3 bar (120 psi)
- 14,1 bar (205 psi)
- 10,3 bar (150 psi)
- 13,4 bar (195 psi)
- 19 bar (275 psi)
- Záruka:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box