Masterflex® Multi-Turn PTFE Needle Valves, Avantor®
Dodavatel: Avantor Fluid Handling
Certifikáty
O tomto produktu
Easily meter corrosive fluids or gases.
- Constructed of durable PTFE
Maximum pressure is 75 psi; maximum temperature is +200 °F (+93 °C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125 inch.
- Environmentally Preferable:
Specifikace
- Catalog No:
- MFLX06393-71
- MFLX06393-68
- MFLX06393-80
- MFLX06393-66
- MFLX06393-82
- MFLX06393-92
- MFLX06393-90
- MFLX06393-94
- MFLX06393-96
- MFLX06393-60
- MFLX06393-61
- MFLX06393-70
- Model:
- 06393-71
- 06393-68
- 06393-80
- 06393-66
- 06393-82
- 06393-92
- 06393-90
- 06393-94
- 06393-96
- 06393-60
- 06393-61
- 06393-70
- Vnitřní Ø potrubí:
- 0,375"
- 0,375"
- 0,375"
- 0,375"
- Vstupní otvor:
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- 3,18 mm (0,125")
- Tubing ext.Ø:
- 0,25"
- 0,25"
- 0,25"
- 0,25"
- Máčené materiály:
- Aluminum, PTFE, CTFE
- Aluminum, PTFE, CTFE
- Stainless Steel, PTFE, CTFE
- Aluminum, PTFE, CTFE
- Stainless Steel, PTFE, CTFE
- Stainless Steel, PTFE, CTFE
- Stainless Steel, PTFE, CTFE
- Stainless Steel, PTFE, CTFE
- Stainless Steel, PTFE, CTFE
- Aluminum, PTFE, CTFE
- Aluminum, PTFE, CTFE
- Aluminum, PTFE, CTFE
- Záruka:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year