Masterflex® Transfer Tubing, Tygon® A-60-F, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Stay compliant with food applications.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
This tubing is ideal for the food, dairy, and beverage industries. Characteristics include heat, ozone, and UV light resistant; non-aging; nonoxidizing; superior acid and alkali resistance. Tubing is manufactured according to GMP and has a temperature range of –60 to 275 °F (–51 to 135 °C). Sterilize with autoclave.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX95710-70
- MFLX95710-90
- MFLX95710-80
- MFLX95710-60
- MFLX95710-50
- MFLX95710-30
- MFLX95710-00
- MFLX95710-10
- MFLX95710-20
- MFLX95710-40
- Model:
- 95710-70
- 95710-90
- 95710-80
- 95710-60
- 95710-50
- 95710-30
- 95710-00
- 95710-10
- 95710-20
- 95710-40
- Klarita:
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- GMP compliant:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- Maximální tlak:
- 0,8 bar (12 psi)
- 0,6 bar (8 psi)
- 0,5 bar (7 psi)
- 0,9 bar (13 psi)
- 0,6 bar (8 psi)
- 0,7 bar (10 psi)
- 2,3 bar (34 psi)
- 1,3 bar (19 psi)
- 0,9 bar (13 psi)
- 1,3 bar (19 psi)
- Maximální teplota:
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- Minimální teplota:
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- Regulatory approvals:
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- FDA;NSF-51
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Pracovní tlak:
- 0,8 bar (12 psi)
- 0,6 bar (8 psi)
- 0,5 bar (7 psi)
- 0,9 bar (13 psi)
- 0,6 bar (8 psi)
- 0,7 bar (10 psi)
- 2,3 bar (34 psi)
- 1,3 bar (19 psi)
- 0,9 bar (13 psi)
- 1,3 bar (19 psi)
- Záruka:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year