10X Tris buffered saline, liquid, Corning®
Catalog # MDTC46-012-CM
Supplier: Corning
CAS Number:
Specifications
- Bal.:6
- Popis:10X tris buffered saline, liquid, 1L
- Skladovací teplota:Ambient
- Rozměry:1 L
- Transportní teplota:Ambient
Specifications
About this item
Tris Buffered Saline (TBS) is an isotonic, non-toxic buffer used in some biomedical techniques to maintain pH within a relatively narrow range.
TBS can shift pH relatively dramatically as temperature changes, with the pKa declining approximately 0,03 units per degree Celsius rise in temperature.