PipeCheck standards for checking volumes of pipettes by photometry, Spectroquant®, Supelco®
Catalog # 1.14962.0001
Supplier: Merck
CAS Number:
Specifications
- Bal.:25 Tests
- Popis:Check solutions (4x6 cells) and reference solutions (4 cells ) for checking the volume of pipettes Spectroquant®
- M-Clarity quality level:MQ100