Hygromycin B solution 50 mg/ml, sterile
Catalog # MDTC30-240-CR
Supplier: Corning
CAS Number:
Specifications
- Bal.:20 mL
- Sekvence:2305
Specifications
About this item
:
:527.5 g/mol
Storage Temperature:Refrigerator
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479