CultiControl freeze-dried microorganism strains derived from NCTC® cultures
Katalogové číslo 70121.
Dodavatel: LIOFILCHEM
Číslo CAS:
Specifikace
- Bal.:5
- Popis:Serratia marcescens derived from NCTC 13382
- Sekvence:89
Specifikace
O tomto produktu
Lyophilised reference stock culture preparations containing a single strain of a microorganism for quality control of culture media, education and proficiency programs. Lyophilisation is a method for long-term preservation of microorganisms. The microorganism preparations are derived from NCTC® (National Collection of Type Cultures by the UK Health Security Agency - UKHSA).
- One single passage from the reference NCTC® culture
- Two years shelf life
- More than 100 freeze-dried microorganisms available
Packaging: CultiControl microorganisms are packaged in a resealable vial that contains five (5) lyophilised pellets of a single microorganism strain, and a desiccant to prevent adverse accumulations of moisture.
Caution: These products are for in-vitro use only.