Tamper-evident closure for compact jerrycan
Katalogové číslo 215-4530
Dodavatel: BURKLE
Specifikace
- Thread:DIN 50
- Barva:Modrá
- Popis:Tamper-evident closure for compact jerrycan
- Materiál:PP/PE
- Environmentally Preferable:
- Bal.:1
Specifikace
O tomto produktu
Screw-on closure with tamper-evident ring for compact jerrycan.
- Screw-on cover made of PP
- Seal made out of PE
- Thread DIN 50
The closure offers protection against tampering and a guarantee that the container has not been opened before, since the snap ring is visibly broken the first time the container is opened.