Orders LinkContactUsLinkComponent
Safety faceshields, Shape™
02, Shape™ safety faceshield, clear visor
Katalogové číslo 111-2594
Dodavatel:   JSP
Safety faceshields, Shape™
Katalogové číslo 111-2594
Dodavatel:  JSP
Safety faceshields, Shape™
Safety faceshields, Shape™
02, Shape™ safety faceshield, clear visor
Přejít na:
Price
531,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
A compact, comfortable, and lightweight faceshield with superior optical quality.
  • Perfect optical quality
  • 2,2 mm polycarbonate visor
  • Wide field of vision
  • Adjustable wheel ratchet headband
  • Compatible with prescription glasses
The Shape™ Safety Faceshield features a moulded polycarbonate visor that provides superior optical quality with an exceptional field of vision.
Certifications: EN 166; EN 170.
Documents
Specifikace
  • Popis:
    Shape™ safety faceshield, clear visor
  • Sekvence:
    02