Clear safety spectacles, Crackerjack™
Katalogové číslo JSPA1CRAK23C
Dodavatel: JSP
Specifikace
- Popis:Crackerjack™ safety spectacles
- Sekvence:01
- Environmentally Preferable:
- Lenses:Clear
- Bal.:1
Specifikace
O tomto produktu
Stylish, single lens, lightweight safety spectacle.
- Ultra enveloping
- Lateral protection
- Flat temples
The Crackerjack™ is a stylish single lens spectacle that is lightweight with flat temples providing comfort for extended periods of use. Its design makes it a perfect fit for wearing in conjunction with helmets and ear defenders.
Certifications: Meets EN 166:2001.T9:V9