Elbow fitting, b.safe
Elbow fitting, PE, GL 25 (male), GL 25 (female)
Katalogové číslo BOHLR1224-01
Dodavatel: Bohlender
Přejít na:
Product Details & Documents
Screw cap for thread GL 25 made of PPS with male thread GL 25.
- Made of static dissipative PE-EX
- With rotary elbow fitting
- Without sagging
Angle of approx. 90° for a space-saving connection of b.safe Waste Tubes to b.safe Desktop Mounts or b.safe Drain Basin. Rotary elbow fitting for freely adapting the direction of the tube connection.
Specifikace
- Závit:GL 25 (male), GL 25 (female)
- Popis:Elbow fitting
- Materiál:PE