Orders LinkContactUsLinkComponent
ISO warning labels
ISO warning labels
Dodavatel:   GA
ISO warning labels
Dodavatel:  GA
Certifikáty
ISO warning labels
ISO warning labels
3 Dostupných variant níže
Product Details & Documents
Pre-printed general warning labels for the identification of equipment, containers, and material that may pose a safety risk in the laboratory environment.
  • Withstand high-temperature conditions up to 100° C (212° F)
  • Pre-printed with full color, yellow on silver background, design
  • Durable thermoplastic material is resistant to wear and tear, water, mild chemicals, and detergents
  • Resistant to gamma irradiation (tested to up to 50kGy)
Coated with a glove-friendly adhesive, these waterproof thermoplastic labels are perfect for labeling water baths, heat blocks, autoclave containers, and instruments or tools with moving parts.
Product Family Options
Informace o produktu
  • Sekvence
    Barva
    Rozměry
    Balené
    Dostupnost
    Price
  • GAINWL-008-0.25R
    ISO warning label - Hot Surface Hazard
    ISO warning label - Hot Surface Hazard
    01
    Černá/žlutá
    19,05×19,05 mm
    250 labels/roll
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 Roll
    1 * 1 Roll - 1 220,00 Kč
    1 220,00 Kč
    1 * 1 Roll
       1 220,00 Kč
  • GAINWL-010-0.25R
    ISO warning label - Low Temperature
    ISO warning label - Low Temperature
    02
    Černá/žlutá
    19,05×19,05 mm
    250 labels/roll
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 Roll
    1 * 1 Roll - 1 220,00 Kč
    1 220,00 Kč
    1 * 1 Roll
       1 220,00 Kč
  • GAINWL-009-0.25R
    ISO warning label - General Warning Symbol
    ISO warning label - General Warning Symbol
    03
    Černá/žlutá
    19,05×19,05 mm
    250 labels/roll
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 Roll
    1 * 1 Roll - 1 220,00 Kč
    1 220,00 Kč
    1 * 1 Roll
       1 220,00 Kč