Orders LinkContactUsLinkComponent
Nailsaver sponge, Scotch-Brite™
Oranžová/zelená, 150×95 mm
Katalogové číslo MMMASCH2282
Dodavatel:   3M
Nailsaver sponge, Scotch-Brite™
Katalogové číslo MMMASCH2282
Dodavatel:  3M
Nailsaver sponge, Scotch-Brite™
Nailsaver sponge, Scotch-Brite™
Oranžová/zelená, 150×95 mm
Přejít na:
Price
4 630,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
Scotch-Brite™ Nailsaver sponge's unique side grooves protect the fingernails while allowing for a better grip for more cleaning leverage.
  • Cleaning sponge with a unique ridge to protect fingernails
  • Grooved pad protects nails, reducing the need for gloves
  • Handy size for a good grip and excellent durability
  • Delicate non-scratch cleaning sponge for bathrooms surfaces
Clean bathrooms without damaging fingernails with Scotch-Brite™ Nailsaver sponge.
This non-scratch cleaning sponge offers good grip and excellent durability. It features unique side grooves that protect the fingernails. This sponge offers worry-free cleaning, reducing the need for gloves and the likelihood of broken nails.
Specifikace
  • Barva:
    Oranžová/zelená
  • D׊:
    150×95 mm