ContactUsLinkComponent
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Catalog # HEWL01018-60025
Supplier:  Agilent
undefined
CrossLab Bottle Head Assemblies for HPLC
Catalog # HEWL01018-60025
Supplier:  Agilent
Specifications
  • Popis:
    Solvent inlet filter, stainless steel
  • Sekvence:
    24
  • Bal.:
    1
Specifications
About this item
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.