Wire weight, OIML F1
Katalogové číslo SARBYCW1131-00
Dodavatel: Sartorius Balances
Specifikace
- Hmotnost:10 mg
- Popis:Wire weight, OIML F1
- Bal.:1
- Poč. závaží:1
Specifikace
O tomto produktu
Accurate metrological weight with nominal mass value of 10 mg.
- OIML class F1
- Hardness: ca. 110 HB
- Density of 7,950 kg/m³
- High gloss polished surface
- No laser markings
- Without certificate
Certifications: Weights comply with the International Recommendation OIML-R111.