Microscope slides, Assistent®
1 mm, 25×25 mm
Catalog # HECH91240000252
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Selected proven glass quality.
- Cleaned
- Ready for use
Specifications
- D׊:25×25 mm
- Tloušťka:1 mm
Selected proven glass quality.