GC columns, J&W CycloSil-B
Katalogové číslo HEWL112-6632LTM
Dodavatel: Agilent
Specifikace
- Film thickness:0,25 µm
- Ø int.:0,25 mm
- Length:30 m
- Sekvence:02
- Environmentally Preferable:
- Fáze:CycloSil-B
- Bal.:1
Specifikace
O tomto produktu
CycloSil-B delivers improved resolution of many chiral separations and is ideal for a number of chiral γ-lactones and terpenes.
- 30% hepatkis (2,3-di-O-methyl-6-O-t-butyl dimethylsilyl)-ß-cyclodextrin in DB-1701
- Chiral separations without chiral specific derivatisation
- Each J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
CycloSil-B GC columns are not bonded or cross-linked and so they does not recommend solvent rinsing.