Stabiliser for proteins and standards, SeramunStab® Stpur
Catalog # APOSBIL3118-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Bal.:100 mL
- Popis:SeramunStab® Stpur stabiliser for proteins and standards
- Sekvence:01
Specifications
About this item
Liquid product colourless to slightly yellow or greenish odourless pH 7,3 to 7,5 boiling point 104 °C. Stabiliser for controls and protein standards.