- Type:Anti-scratch
- Barva:Průhledná
- Popis:Lens marking: 2C-1.2 3M 1 FT
- Environmentally Preferable:
- Lenses:Clear PC
- Bal.:1
The SecureFit™ 200 Series are lightweight safety glasses with 3M™ pressure diffusion temple technology. These glasses feature a wraparound design that's secure and comfortable. They are available in various lens tints.
- Economical lightweight (18 g) safety glasses
- Innovative SecureFit™ pressure diffusion temple technology design features slotted ribs that provide comfortable pressure equalisation
- Temples flex and self-adjust to the size of wearer's head
- Flat and flexible temples combine easily with earmuffs
- Easy to clean
- Foam gasket available to help limit eye exposure to debris
- Prescription insert available
For lightweight comfort at an affordable price, these safety glasses weigh just 18 g and offer a comfortable fit. The Pressure Diffusion Temple Technology allow the temple touchpoints to naturally adjust to an individual's head size. Wide flexible temple arms over the ears provide solid side protection and work well with ear muffs. The wraparound design gives the user a wide field of vision and helps to reduce slippage from head movements.
Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties (available on selected models). Wraparound design improves the security of fit and prevents slipping.