ContactUsLinkComponent
Intuvo gas clean filter kit
  HEWLCP17995
 :  Agilent
Intuvo gas clean filter kit
  HEWLCP17995
 :  Agilent
  • Popis:
    Gas clean filter kit, Intuvo, includes bracket, connecting unit, and filter
  • Bal.:
    1 KIT
This Gas Clean Filter System delivers clean gases, reducing the risks of column damage, sensitivity loss, and instrument downtime.
  • Deliver clean gases for accurate analyses
  • Fast, leak-free filter replacement reduces downtime
  • Highly sensitive filter indicators provide maximum instrument protection
Inserting a Gas Clean Filter System in the gas line immediately before the instrument inlet greatly reduces the level of impurities, thus improving trace analysis. Contaminants entering your GC column will also be reduced, which is critical for high-temperature analysis and essential for longer column life.
 : Includes bracket, connecting unit and carrier.