RF work/load coil for PerkinElmer
HEWL8003-0559
: Agilent
- Popis:RF load coil assembly, for NexION/ELAN 9000/6×00/DRCs
- Bal.:1
These coils are equivalent to PerkinElmer ICP-MS coils for complete confidence.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage