GS-Alumina KCl GC columns, J&W
HEWL115-3352E
: Agilent
- Ø vnitř.:0,53 mm
- Délka:50 m
- Sekvence:02
- Fáze:GS-Alumina KCI
- Bal.:1
GS-Alumina KCI is the least polar PLOT alumina phase and is a good choice for light hydrocarbon analysis.
- Least polar Alumina phase
- Aluminum oxide deactivated with KCl
- Good choice for light hydrocarbon analysis
- Good resolution of propadiene and butadiene from ethylene and propylene streams
- Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
Agilent JW GS-Alumina KCI is the least polar PLOT alumina phase and is made from aluminum oxide, with proprietary deactivation. GS Alumina KCI is a good choice for light hydrocarbon analysis.