GC Columns, J&W CP-Sil 43 CB
Catalog # VARICP7745
Supplier: VARIAN
Specifications
- Tloušťka filmu:0,20 µm
- Ø vnitř.:0,32 mm
- Délka:25 m
- Fáze:CP-Sil 43 CB
- Bal.:1
Specifications
About this item
CP-Sil 43 CB separates aromatic from aliphatic hydrocarbons with moderate polarity for specific selectivity equivalent to OV-255.
- 25% cyanopropyl/25% phenyl/50% dimethylpolysiloxane phase
- Mid polarity
- Solvent rinsable
Agilent JW CP-Sil 43 CB is a chemically bonded and cross-linked, moderately polar column.