Multiple affinity removal spin cartridge reagent kit
Catalog # AGLS5188-5254
Supplier: AGILENT
CAS Number:
Specifications
- Bal.:1 kt
- Popis:Multiple affinity removal spin cartridge reagent kit
- Sekvence:02
Specifications
About this item
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Delivery: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.