GC columns, DB-35ms, J&W
HEWL122-3832LTM
: Agilent
- Tloušťka filmu:0,25 µm
- Ø vnitř.:0,25 mm
- Délka:30 m
- Sekvence:03
- Fáze:DB-35ms
- Bal.:1
DB-35ms offers excellent inertness for active compounds while providing very low-bleed characteristics. They are ideal for confirmational analyses.
- Virtually equivalent to (35% Phenyl) methylpolysiloxane
- Mid polarity with very low bleed characteristics, ideal for GC/MS
- Extended temperature limit of 340/360 °C
- Excellent inertness for active compounds
- Ideal for confirmational analyses
J&W DB-35ms is precision-engineered to deliver excellent performance in a range of GCMS applications, particularly in confirmational analyses. These mid-polarity columns operate over an extended temperature range up to 340/360 °C. The bonded and cross-linked DB-35ms columns also feature very-low-bleed characteristics and excellent inertness and so they are particularly suited for analysis of active compounds.