Orders LinkContactUsLinkComponent
Hermetic Tamper Evident Buckets, Rectangular
Hermetic Tamper Evident Buckets, Rectangular
Dodavatel:   LAMAPLAST
Hermetic Tamper Evident Buckets, Rectangular
Dodavatel:  LAMAPLAST
Certifikáty
Hermetic Tamper Evident Buckets, Rectangular
Hermetic Tamper Evident Buckets, Rectangular
3 Dostupných variant níže
Product Details & Documents
Robust PP bucket with ergonomic handle. It features pressure lid with opening-off seal.
  • Rectangular shape
  • Flat bottom ensures total emptying and easy cleaning
  • Especially suitable in cosmetic, pharma, paint or dye industries
  • Food contact material approved
Hot filling possible up to 95 °C.
Product Family Options
Informace o produktu
  • Sekvence
    Barva
    Kapacita
    Výška
    Dostupnost
    Price
  • LAMA1105
    Rectangular bucket with plastic handle, 243×168 mm (top), 218×143 mm (base)
    Rectangular bucket with plastic handle, 243×168 mm (top), 218×143 mm (base)
    01
    Bílá
    3 L
    119 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 175,00 Kč
    175,00 Kč
    1 * 1 KS
       175,00 Kč
  • LAMA1107
    Rectangular bucket with plastic handle, 286×198 mm (top), 255×168 mm (base)
    Rectangular bucket with plastic handle, 286×198 mm (top), 255×168 mm (base)
    02
    Bílá
    5 L
    149 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 222,00 Kč
    222,00 Kč
    1 * 1 KS
       222,00 Kč
  • LAMA1109
    Rectangular bucket with plastic handle, 365×247 mm (top), 332×210 mm (base)
    Rectangular bucket with plastic handle, 365×247 mm (top), 332×210 mm (base)
    03
    Bílá
    11 L
    191 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 398,00 Kč
    398,00 Kč
    1 * 1 KS
       398,00 Kč