Ceramic Homogenisers
HEWL5982-9313
: Agilent
- Popis:QuEChERS extract tubes, AOAC Method with ceramic homogeniser
- Sekvence:03
- Bal.:100
QuEChERS ceramic homogenisers increase lab productivity by reducing extraction time while maintaining consistency.
- Cutting the required extraction time from 60 seconds to as little as 20 seconds – a time savings of 70% per sample
- Maintaining high, reproducible extractions in a third of the time
- Minimizing variance between technicians
- Breaking up salt agglomerates and maintaining a consistent grinding of homogenising material
The same great ceramic homogenisers available in our QuEChERS Kits are also available to buy in bulk, providing excellent grinding capabilities of the samples.