GC columns, J&W DB-XLB
Katalogové číslo HEWL123-1236
Dodavatel: Agilent
Specifikace
- Tloušťka filmu:0,50 µm
- Ø vnitř.:0,32 mm
- Délka:30 m
- Sekvence:11
- Fáze:DB-XLB
- Bal.:1
Specifikace
O tomto produktu
DB-XLB is exceptionally low bleed for active compounds, specifically pesticides, herbicides, PCBs, and PAHs, and for confirmation analyses.
- Exceptionally low bleed and low polarity; unique selectivity
- Excellent inertness for active compounds
- Ideal for confirmational analyses
- Excellent for pesticides, herbicides, PCBs and PAHs
- Ideal for GC/MS; MSD testing and certification available
The column delivers highly specific analysis of pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). DB-XLB is also excellent as a confirmation column for semivolatile environmental analyses.