ContactUsLinkComponent
Desalting-RP HPLC columns, AdvanceBio
Desalting-RP HPLC columns, AdvanceBio
Katalogové číslo HEWLPL1612-1102
Dodavatel:  Agilent
undefined
Desalting-RP HPLC columns, AdvanceBio
Katalogové číslo HEWLPL1612-1102
Dodavatel:  Agilent
Specifikace
  • Formát kolony:
    Guard cartridge
  • Ø vnitř.:
    2,10 mm
  • Délka:
    12,5 mm
  • Bal.:
    3
Specifikace
O tomto produktu
The key features and benefits of the AdvanceBio Desalting RP columns.
  • Allow MS detection with traditionally non-MS friendly techniques such as IEX, SEC, and affinity chromatography
  • Save time by desalting your samples on-line
  • Concentrate samples prior to elution
  • Reduce down time and maintenance by keeping your MS cleaner
  • Improve overall data confidence by improving the quality of your data
Reversed-phase desalting for on-line removal of salt ions before MS detection affinity, ion exchange, and size exclusion chromatography are commonly used techniques in the analysis of proteins such as monoclonal antibodies. However, these techniques require aqueous mobile phases that contain non-volatile salts. These non-volatile salts present a problem when using MS detection as they cause signal suppression and can contaminate the MS detector through salt deposition leading to increased maintenance and instrument downtime. AdvanceBio Desalting-RP cartridges solve this problem, enabling fast and efficient on-line removal of salt ions before MS detection. These cartridge style columns can be used on their own on any LC system to desalt collected fractions. Or on an 1290 Infinity II 2D-LC system as the second dimension to desalt after the first dimension separation.