Recombinant Cytomegalo Virus pP28 (UL99)
Catalog # APOSBITP1840-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Bal.:1 mg
- Typ proteinu/peptidu:Recombinant
- Proteinová synonyma:CMV Pp28
- Název proteinu/peptidu:Cytomegalo Virus pP28 (UL99)