ContactUsLinkComponent
Recombinant Hepatitis B Virus Core (1-186)
Recombinant Hepatitis B Virus Core (1-186)
Catalog # APOSBITP1820-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis B Virus Core (1-186)
Catalog # APOSBITP1820-1MG
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Bal.:
    1 mg
  • Typ proteinu/peptidu:
    Recombinant
  • Proteinová synonyma:
    HBcAg
  • Název proteinu/peptidu:
    Hepatitis B Virus Core (1-186)