0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteiny
Recombinant Chlamydia Trachomatis PGP3-D (from CHO cells)
Recombinant Chlamydia Trachomatis PGP3-D (from CHO cells)
APOSBITP1742-1MG
:
Apollo Scientific
:
Recombinant Chlamydia Trachomatis PGP3-D (from CHO cells)
APOSBITP1742-1MG
:
Apollo Scientific
:
Bal.:
1 mg
Typ proteinu/peptidu:
Recombinant
Zdroj:
CHO cells
Proteinová synonyma:
Chlamydia PGP-3D
Název proteinu/peptidu:
Chlamydia Trachomatis PGP3-D