Order Entry
Czech Republic
ContactUsLinkComponent
 
Recombinant Herpes Simplex Virus-1 gG
Catalog # APOSBITP1737-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Herpes Simplex Virus-1 gG
Catalog # APOSBITP1737-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Bal.:
    1 mg
  • Typ proteinu/peptidu:
    Recombinant
  • Proteinová synonyma:
    HSV-1 gG
  • Název proteinu/peptidu:
    Herpes Simplex Virus-1 gG

Specifications

About this item