0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteiny
Human Recombinant Creatine Kinase MB Isoenzyme Type-1
Human Recombinant Creatine Kinase MB Isoenzyme Type-1
APOSBITP1434-1MG
:
Apollo Scientific
:
Human Recombinant Creatine Kinase MB Isoenzyme Type-1
APOSBITP1434-1MG
:
Apollo Scientific
:
Bal.:
1 mg
Typ proteinu/peptidu:
Recombinant
Druhy:
Human
Proteinová synonyma:
CKMB Type-2
Název proteinu/peptidu:
Creatine Kinase MB Isoenzyme Type-1