0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteiny
Mouse recombinant MIP-1a
Mouse recombinant MIP-1a
APOSBITC2035-1MG
:
Apollo Scientific
:
Mouse recombinant MIP-1a
APOSBITC2035-1MG
:
Apollo Scientific
:
Bal.:
1 mg
Typ proteinu/peptidu:
Recombinant
Druhy:
Mouse
Proteinová synonyma:
Macrophage Inflammatory protein-1 alpha, CCL3
Název proteinu/peptidu:
MIP-1a