Cathepsin inhibitor I
APOSBIC1020-1MG
:
- Bal.:1 mg
- Proteinová synonyma:Z-Phe-Gly-NHO-Bz
- Název proteinu/peptidu:Cathepsin Inhibitor I
Selectively inhibits Cathepsin B, Cathepsin L, Cathepsin S and Papain.
Selectively inhibits Cathepsin B, Cathepsin L, Cathepsin S and Papain.