Deep well plate, 96-well, WHEATON® μLplate®
Catalog # WHEA07-2000
Supplier: DWK Life Sciences
Specifications
- Sterilní:-
- Barva:Průhledná
- Popis:μLplate® 96-well PP microplate, round bottom, round well
- Environmentally Preferable:
- Bal.:1
- Well volume:500 µl
Specifications
About this item
500 μl round well, round bottom, 96-well microplate.
This 96-well round well microplate is made of polypropylene and available in a variety of volumes. Microplates are manufactured to ANSI specifications to guarantee a smooth integration into the automated work flow. Round and square well covers are available separately as pierceable, standard, and pre-cut. Covers manufactured of Ethylene Vinyl Acetate (EVA) are pierceable by standard needle configurations and have a superior seal to similar products. For those needing reseal ability, the available silicone mats with a spayed-on PFTE barrier are available as standard or pre-cut.
Certifications: ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004.
Ordering information: Cover not included.