Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-(4-Amino-2-(trifluoromethyl)phenyl)piperazin-1-yl)ethanone
Irritant
1-(4-(4-Amino-2-(trifluoromethyl)phenyl)piperazin-1-yl)ethanone
Catalog # APOSPC8825-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-(4-Amino-2-(trifluoromethyl)phenyl)piperazin-1-yl)ethanone
Catalog # APOSPC8825-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Refrigerator
CAS Number:914348-88-2
Formula:C₁₃H₁₆F₃N₃O