Order Entry
Czech Republic
ContactUsLinkComponent
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride 97%
Corrosive
Irritant
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride 97%
Catalog # APOSPC49253-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride 97%
Catalog # APOSPC49253-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    100 g
  • Sekvence:
    3401

Specifications

About this item

 :
 :276.08 g/mol
Melting Pt:208…214 °C
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
CAS Number:127337-60-4
Formula:C₉H₉ClF₃NO
MDL Number:MFCD00800224