Order Entry
Czech Republic
ContactUsLinkComponent
[5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazolyl]methanol 97%
[5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazolyl]methanol 97%
Catalog # APOSPC4922-500MG
Supplier:  Apollo Scientific
CAS Number:  
[5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazolyl]methanol 97%
Catalog # APOSPC4922-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    500 mg
  • Sekvence:
    2725

Specifications

About this item

 :
 :309.68 g/mol
Melting Pt:112…114 °C
Storage Temperature:Ambient
CAS Number:338410-31-4
Formula:C₁₁H₁₁ClF₃N₃O₂
MDL Number:MFCD00202658